Mini Columns, Caplugs
Supplier: Caplugs
Jump to:
À propos de cet article
Mini Columns are molded in polypropylene for molecular biology studies.
- 0.5 ml mini-column is 4.83 cm (1.9") overall in height
- 2.4 ml mini-column is 3.81 cm (1.5") overall in height
Columns feature a HDPE filter of medium porosity (45 to 90 µm)
3 Options Available Below
Product Details & Documents
Mini Columns are molded in polypropylene for molecular biology studies.
- 0.5 ml mini-column is 4.83 cm (1.9") overall in height
- 2.4 ml mini-column is 3.81 cm (1.5") overall in height
Columns feature a HDPE filter of medium porosity (45 to 90 µm)
Product Family Options
Informations sur le produit
- DésignationColumn HardwareGranulométrieØ int.LongueurCapacitéAvailabilityPrice
- Supplier Number: 208-1289-03S
- Mini columns, naturalPP45 - 90 µm38.30 mm16 mm2.4 mlLog in pour la disponibilitéCarton of 1,000$412.59Supplier Number: 208-3049-030
- Mini columns, naturalPP45 - 90 µm38.30 mm16 mm2.4 mlLog in pour la disponibilitéCarton of 200$134.88Supplier Number: 208-3049-03S