Extraction Columns, Caplugs
Extraction columns, 3 - ³/₈", medium filter, PP, 45 - 90 µm
Catalog # 76498-812
Supplier: Caplugs
Fournisseur #: 208-3355-030
Jump to:
Product Details & Documents
Extraction columns are for use in drug extractions, HPLC sample preparation, general chromatography applications and other research procedures.
- Columns measure 3 to ³/₈" long
- Include a medium (45 to 90 μm) polyethylene filter matrix support
- Large reservoirs and cartridge portions with straight sidewalls accommodate solid-phase packing material such as silica and alumina
Columns are available in either natural polypropylene or light-shielding amber color.
Spécifications
- Granulométrie:45 - 90 µm
- Désignation:Extraction columns, 3 - ³/₈", medium filter
- Column Hardware:PP