Sera- Separa Columns, Caplugs
Supplier:   Caplugs
Sera- Separa Columns, Caplugs
Supplier:  Caplugs
Certificates
Sera- Separa Columns, Caplugs
Sera- Separa Columns, Caplugs
2 Options Available Below
Product Details & Documents
Sera-Separa columns are ideal for use in removing fibrin and macro particles from serum to prevent line clogging.
  • Graduated barrel tells exactly how much liquid has been filtered
  • Wide-body style prevents sample probes from hitting side walls
Users insert the 16 mm flange into the evacuated blood tube and push downward.
Product Family Options
Informations sur le produit
  • Désignation
    Column Hardware
    Capacité
    Availability
    Price
  • Sera-Separa columns, 2.625"
    PE
    5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Carton of 1,000
    Carton of 1,000 - $978.82
    $978.82
    Carton of 1,000
       $978.82
    Supplier Number: 208-3193-020
  • Sera-Separa columns, 4.25"
    PE
    9 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Carton of 1,000
    Carton of 1,000 - $1,045.58
    $1,045.58
    Carton of 1,000
       $1,045.58
    Supplier Number: 208-3084-020