ESD Safe Microscope Covers, Transforming Technologies
ESD safe microscope cover, 10.25×6.5"
Catalog # 76474-668
Supplier: Transforming Technologies
Supplier #: MC1221W
ESD Safe Microscope Covers, Transforming Technologies
Catalog # 76474-668
Supplier: Transforming Technologies
Jump to:
Product Details & Documents
Protect microscope optics from dust and other contamination while keeping the work station ESD safe.
- Permanently anti-static: less than 10⁶
- Made with polyester and conductive yarn
- Fits most common microscopes
Common microscope covers are made from vinyl, a material that is an insulator and potential static generator. This cover is made from ESD safe materials and does not generate a significant charge.
Spécifications
- Désignation:ESD safe microscope cover
- Taille:10.25×6.5"