Spécifications
- Color:Black
- Description:ESD heel grounder for large footwear
- Resistor:1 MΩ
- Cat. no.:76474-662
Spécifications
À propos de cet article
Ground mobile personnel with large size footwear with long-lasting stretch Hook and Loop ESD heel grounders.
- Reversible cup design allows for longer life
- Stretch hook and loop strap creates a secure and comfortable fit around the shoe
- Buried 1 meg resistor for safety
This Heel Grounder is constructed from durable materials and features a stretch hook and loop closure strap to maximize fit and comfort. The cup is designed to fit larger sized footwear, such as men’s sizes 12 and above. The cup of the device is a symmetrical design that can be reversed when put on to double the lifespan. The device connects the person wearing them to ground via a proper floor mat or flooring material. Wearing the conductive ribbon inside the shoe or sock assures proper electrical contact with the user. The ribbon connects to the conductive rubber to complete the circuit between the operator and the static controlled floor or mat.
A buried 1 meg ohm resistor ensures safety and is standard. These heel grounders must be worn on both feet to provide consistent grounding while in motion.