Ptec™ Self-Cleaning Bench Top Ionizer, Transforming Technologies
Catalog # 76474-666
Supplier: Transforming Technologies
Specifications
- Description:Self-cleaning bench top ionizer
- Cat. no.:76474-666
Specifications
About this item
The Ptec™ IN5500 AC benchtop air ionizer is a small, efficient static elimination device with variable fan speed for protecting electronic components and general static protection.
- Remove static charges quickly and efficiently
- Self-cleaning system simplifies service and maintenance
- Perfect for single station table top, bench, or desk use
- Balance: ±10 V
- Small footprint
The unit can self-clean, which eliminates maintenance costs and equipment downtime and ensures the ionizer provides optimal static eliminating performance. Using AC corona technology to make the air more conductive, Ptec™ ionizers produce a balanced stream of positive and negative air ions that never need calibration. These devices are used in many industries and manufacturing applications where static electricity is a problem.