Warmbier Veribox ESD Verification Resistor Box, Transforming Technologies
Catalogue # 76474-632
Fournisseur: Transforming Technologies
Spécifications
- Description:Veribox ESD verification resistor box
- Cat. no.:76474-632
Spécifications
À propos de cet article
Verify that ESD testing equipment is operating correctly with the Warmbier Veribox.
- For the periodic verification of ESD testers
- Contains fixed resistors: 1GΩ, 1MΩ, 10KΩ
- Passive device; uses no power
- For use with most available resistance meters
The Veribox is designed for the periodic verification of ESD test equipment such as Surface Resistance Meters. The Veribox contains fixed resistors that can quickly and accurately simulate resistance for testing the accuracy of ESD testers and meters.