Warmbier SRM200/EFM51 ESD Testing Starter Kit, Transforming Technologies
Catalog # 76474-628
Supplier: Transforming Technologies
Specifications
- Description:SRM200/EFM51 ESD testing starter kit
- Cat. no.:76474-628
Specifications
About this item
A kit to test basic ESD measurements such as surface resistivity and electrostatic fields.
A complete and very useful tool kit for testing and measuring different products and materials within an electrostatic protected area (EPA) such as bags, clothes, flooring, chairs, tabletops, boxes.
Delivery: The kit consists of a EFM51 electrostatic field meter and one SRM200 surface resistance meters and is supplied in a conductive case.