Cleanroom Safe ESD Polyester Wipe, WP7200, Transforming Technologies
Catalog # 76474-832
Supplier: Transforming Technologies
Specifications
- Description:Cleanroom safe ESD polyester wipe
- Cat. no.:76474-832
- W×L:22.9×22.9 cm (9×9")
Specifications
About this item
Ideal for wiping exterior areas of products in ESD-sensitive areas as well as cleaning of process tools and other equipment.
- 100% continuous polyester filament with conductive carbon yarn
- Good solvent resistance
- Lint-free and particle-free
- 150 wipes per bag
Made of 100% continuous polyester filament with conductive yarns designed to dissipate electrical charge and control ESD. Wipes are static dissipative on both sides, unlike similar wipers that are dissipative on only one side. The edges are laser sealed to ensure ultra-low lint and particle generation. Suitable for use in Class 100-1000 cleanrooms.