Elastic Fabric Adjustable Wrist Strap Sets, WB8000 Series, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
ESD wrist strap set is a comfortable ESD grounding device for industrial and controlled environments.
- Unique locking clasp eliminates excess band material when tightened
- Soft woven fabric of the wrist band allows the skin to breathe
- Highly conductive copper sulfide thread for conductivity
- Band expands to fit most wrists
- 4 mm snap fits most common coil cords
The WB8000 Series Wrist Band and Coil Cord set features an elastic adjustable band with a unique locking clasp that eliminates excess wrist band material. The band is standard with 4 mm, machined male snap and provides a 360° conductive contact around the wrist, providing maximum skin contact. The Band Set is completed with a premium coil cord featuring 2.5 mm polyurethane coil insulation with excellent coil memory. A shorter, swivel type banana jack increases coil life, prevents cord tangles, and fits most bench grounds. It is uses highly conductive copper sulfide nylon thread for conductivity and it has a maximum resistance of 160 ohms.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20 and ESDA Standard 1.1-2006.
Frequently Bought Together