Spécifications
- Description:Disposable wrist band
- Cat. no.:76474-798
Spécifications
À propos de cet article
The disposable wrist strap provides cost-effective and reliable static control protection for short-term use. Perfect for guests, field service calls, or for shipping with ESD sensitive products.
- Designed for one-time use
- Packaged individually in a printed bag with instructions
- Ready to pack with ESD sensitive products
The Disposable Wrist Strap offers a safe, effective, and economical means for providing static protection to end users of electronic devices. It can be included with shipments of computer upgrades, network systems, and other devices to provide protection at the customer's site. Constructed from a durable non-woven fabric impregnated with a conductive polymer, it provides 360° skin contact to assure proper skin to band resistance. A convenient loop at one end of the device facilitates the formation of a wrist strap. The other end of the device uses a conductive adhesive on a copper foil for easy contact to any electrical ground. A current limiting resistance is built into the fabric for operator safety.