Dual Conductor Metal Wrist Strap Sets, WB5000 Series, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
The adjustable WB5000 series dual conductor wrist strap set is designed for use with a Dual Conductor Constant Workstation Monitor to maximize ESD protection.
- Premium metal expansion wrist strap with more reliable path-to-ground
- Superior contact with skin
- Dual-Wire conductor provides redundancy
- Fixed size bands available in three sizes
Adjustable Dual Wire Wrist Strap sets provide redundancy by including two connections to ground, so even if one dual wire wrist strap conductor is severed, the operator has reliable path-to-ground with other wire. The Speidel metal band features two 180° conductive paths around the wrist, providing maximum skin contact. The expandable band is constructed with two stainless steel back plates and is available in small, medium, and large sizes. A double insulated jacket provides incredible durability and unmatched reliability.
Certifications: Meets requirements of ANSI/ESD S20.20 and ANSI/ESD S1.1.
Frequently Bought Together