Anti-Allergy Wrist Band and Coil Cord Sets, WB5600 Series, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
This Anti-allergy ESD wrist strap set is the ideal grounding device for people with allergies to common metals.
- No metal contacts the skin, unlike common wrist straps that use a metal plate
- Hypoallergenic silver-plated nylon thread for conductivity
- Back plate has a plastic cover so that metal does not meet skin
Anti-allergy wrist band sets feature hypoallergenic silver-plated nylon thread for conductivity and a plastic covered backplate so that metal does not meet skin. The wrist band and coil cord set feature a 360° conductive contact around the wrist, providing maximum skin contact while allowing the skin to breathe. The 4mm snap works in combination with most standard wristbands. A swivel type banana jack increases coil life and prevents cord tangles.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20 and ESDA Standard 1.1-2006.
Frequently Bought Together