Premium Single Wire Wrist Strap and Coil Cord Sets, WB4000 Series, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
The WB4000 Series Wrist Band and Coil Cord set is a premium band set featuring a machined 4 mm snap post.
- Soft woven fabric is comfortable to wear for long periods
- Machined snap provides a more secure connection to the coil cord
- Cord is durable and has excellent coil memory
The soft wrist band features a 360° conductive contact around the wrist, providing maximum skin contact. The band uses hypoallergenic silver-plated nylon thread for conductivity and has a maximum resistance of 160 ohms. The soft, blue woven fabric of the wrist band allows the skin to breathe and is very comfortable to wear. A shorter, swivel type banana jack increases coil life, prevents cord tangles, and fits most bench grounds.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20 and ESDA Standard 1.1-2006.
Frequently Bought Together