Magnetic Dual Conductor Fabric Wrist Band with Coil Cord, WB3000 Series, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
The WB3000 Series is a high-quality, adjustable anti-static wrist strap with a coil cord. The snaps are specially designed magnets which create a superior electrical connection between the wrist band and coil cords.
- Premium adjustable fabric wrist strap and coil cord set
- Superior contact with the skin for reliable grounding
- Dual-Wire conductor provides redundancy
The magnet snaps eliminate the intermittent failures and wear and tear of traditional snap and spring connections. The soft, blue woven fabric wrist band allows the skin to breathe and makes it very comfortable to wear. The band uses hypoallergenic silver-plated nylon thread for conductivity and a non-allergenic stainless steel base plate for excellent skin contact. The black coil cord terminates with a 3.5 mm stereo plug and includes a 1 meg, molded resistor for safety.
Certifications: Meets requirements of ANSI/ESD S20.20 and ANSI/ESD S1.1.
Frequently Bought Together