About this item
A dual-wire wrist strap set provides fail-safe ESD protection because it has two paths-to-ground. If one wire is broken, the second wire will maintain the connection to ground.
- For use with dual wire resistance constant monitors
- High-quality fabric adjustable wrist band is hypoallergenic and comfortable
The adjustable WB2500 series dual wire fabric wrist strap set features two 180° conductive paths around the wrist, providing maximum skin contact. The band is standard with 4mm male snaps and is used with dual wire resistance based constant monitors. Dual wire wrist straps provide redundancy. When used with dual wire resistance monitors, the WB2500 series wrist strap sets continuously verify resistance to ground. The wrist strap is constructed with a stainless-steel back plate, nylon molding, and a premium elastic yarn that provides excellent fit for comfort and long life. The polyurethane coil insulation and strain relief molding offers excellent coil memory and endurance.