Single Wire Wrist Band and Coil Cord Sets, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
These versatile wrist strap and coil cord sets are available with a 7- or 10-mm snap. The set is a durable and long-lasting method of grounding personnel.
- Durable and long-lasting design
- Comfortable to wear for extended periods
- Fabric band uses hypoallergenic silver-plated nylon thread
- One size fit all
This Wrist Band and Coil Cord set is the most versatile and cost-effective band set. The soft, blue woven fabric and 360° curved buckle of the wrist band allows the skin to breathe and makes it very comfortable to wear. The band uses hypoallergenic silver-plated nylon thread for conductivity and has a maximum resistance of 160 ohms. The coil cords 2.5 mm polyurethane coil insulation offers excellent coil memory. A swivel type banana jack increases coil life and prevents cord tangles.
Frequently Bought Together