VinylStat B Three-Layer ESD Table Mat with Foam Back, Transforming Technologies
Fournisseur: Transforming Technologies
Certificats
À propos de cet article
VinylStat B provides excellent ESD protection and extra cushion for component safety and operator comfort.
- Provides extra cushion for component safety and operator comfort
- Three layers of vinyl with excellent ESD protection
- Available in blue or grey; supplied as mat rolls
Table mats utilize a three-layer construction consisting of a conductive metalized fleece sandwiched between a solid, vinyl top surface and a foam, base layer. Surface resistivity is in the dissipative range: 10⁷ to 10⁸.
Certifications: Meets or exceeds requirements of ANSI/ESD S20.20 per ANSI/ESD STM4.1.
Frequently Bought Together