Cole-Parmer® Universal Pipette Tips with Filter, Sterile
Protect against cross-contamination with tips free of human touch.
- Pure porous polyethylene filter protects against cross-contamination
- Durable polypropylene construction
- Noncytotoxic and nonhemolytic
- Sterilized and DNase, RNase, and pyrogen free
Cole-Parmer sterile pipette tips are constructed of high-grade polypropylene and a polyethylene filter which prevents liquids and aerosols from entering the body of your pipette. Tips are 99% Bacterial Aerosol Filtration Efficient (BFE), ASTM F21012, ISO 10993-5 MRM, minimum essential media elution, and GLP compliant.
: Racks (96 tips per rack): These racks, with smooth rounded edges are extremely durable and a snap latch.
Frequently Bought Together