PCR Expansion, Oxford Nanopore Technologies
76487-126
:
- Description:PCR expansion
- Compatible with:R10.4.1 flow cells
- Cat. no.:76487-126
- No. of Reactions:24
PCR Primers (PRM) and PCR Adapters (PCA).
- Additional PCR primers and adapters for use with Ligation Sequencing Kits
This kit contains PCR Primers and PCR Adapters, sufficient for at least 24 reactions, and is compatible for use with the Ligation Sequencing Kits (76487-140 and 76487-138).
: Flow cells and kits are shipped together at 2 to 8 °C. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.
: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.