Ice-Pak™ Foam Bricks, Cryopak Verification Technologies
Fournisseur: Cryopak Verification Technologies
Certificats
À propos de cet article
Vacuum-sealed in durable film, these bricks are ideal for defined and precise packaging configurations.
- Durable materials
- Reusable
- Consistent structure
Ice-Pak™ foam bricks have a freezing/melting point of 0 °C and offer superior thermal performance when transporting temperature-sensitive products. They feature a consistent, uniform structure that is maintained even after freezing.
Frequently Bought Together