Linbro® Plate Sealer, MP Biomedicals
Catalog # IC097740005
Supplier: MP Biomedicals
Specifications
- Optical property:Optically clear
- Description:Plate sealer
- L×W:13.3×8.3 cm
- Material:Acetate
- Cat. no.:IC097740005
- Packaging:Box of 100
Specifications
About this item
Microplate sealing tape used to provide a tight seal on 96-well plates to minimize evaporation and condensation for short and long term storage.
- Easy to apply and remove on all types of microplates
Microplates can be stored in a wide temperature range, from –32 to –93 °C.