Test Tubes, Caplugs
À propos de cet article
Test tubes provide clarity and can be used for mild acidic or alkali solutions, alcohol and salt, with exceptions of oxidizing acids and high concentrate aclohols.
- Provided with or without colors
- Tubes are bulk packed, non-sterile
- Polystyrene tubes withstand up to 8200×g
- Polypropylene tubes withstand up to 10000×g
- 12×75 mm tubes are available with internally ribs for use in antibody coating and/or protein adsorptions, six slanted ribs increase useful surface area
The polypropylene tubes are chemically resistant and are appropriate for use with organic solvents and stronger acids, alkalis, alcohols, and chlorinated and aromatic hydrocarbons.
24 Options Available Below
Product Details & Documents
Test tubes provide clarity and can be used for mild acidic or alkali solutions, alcohol and salt, with exceptions of oxidizing acids and high concentrate aclohols.
- Provided with or without colors
- Tubes are bulk packed, non-sterile
- Polystyrene tubes withstand up to 8200×g
- Polypropylene tubes withstand up to 10000×g
- 12×75 mm tubes are available with internally ribs for use in antibody coating and/or protein adsorptions, six slanted ribs increase useful surface area
The polypropylene tubes are chemically resistant and are appropriate for use with organic solvents and stronger acids, alkalis, alcohols, and chlorinated and aromatic hydrocarbons.
Product Family Options
Tubes without caps
Informations sur le produit
- CapacitéMatièreColorDiameter ×HCap ColorCap TypeBottom StyleAvailabilityPrice
- Supplier Number: 214-2001-010
- Supplier Number: 214-2002-010
- Supplier Number: 214-2009-010
- Supplier Number: 214-2003-010
- Supplier Number: 214-2023-010
- Supplier Number: 214-2131-010
- Supplier Number: 214-2012-010
- Supplier Number: 214-2030-010
- Supplier Number: 214-2070-010
- Supplier Number: 214-2065-01N
- Supplier Number: 214-2097-010
- Supplier Number: 214-2309-030
- Supplier Number: 214-2355-030
- Supplier Number: 214-2367-030
- Supplier Number: 214-2367-B30
- Supplier Number: 214-2367-R30
- Supplier Number: 214-2367-Y30
- Supplier Number: 214-2313-030
- Supplier Number: 214-2380-030
- Supplier Number: 214-2345-03N
Tubes with caps
Informations sur le produit
- CapacitéMatièreColorDiameter ×HCap ColorCap TypeBottom StyleAvailabilityPrice
- 8 mlPSNatural13×100 mmGreenPE screw capRound bottomLog in pour la disponibilitéCarton of 1,000$358.11Supplier Number: 214-2068-G50
- 8 mlPPNatural13×100 mmGreenPE screw capRound bottomLog in pour la disponibilitéCarton of 1,000$300.53Supplier Number: 214-2079-G60
- 19 mlPSNatural16×125 mmGreenPE screw capRound bottomLog in pour la disponibilitéCarton of 1,000$311.15Supplier Number: 214-2088-G50
- 21 mlPSNatural16×150 mmGreenPE screw capRound bottomLog in pour la disponibilitéCarton of 1,000$418.18Supplier Number: 214-2099-G50