CF® Dye Hydrazide, Biotium
CF® Dye Hydrazide, Biotium
 :   Biotium
CF® Dye Hydrazide, Biotium
 :  Biotium
CF® Dye Hydrazide, Biotium
CF® Dye Hydrazide, Biotium
Fluorescent CF® Dye hydrazides are bright, extremely water-soluble, nontoxic molecules, useful as aldehyde-fixable cell tracers. They can also be used to label aldehyde or ketone groups of polysaccharides, carbohydrates or glycoproteins.
  • Fluorescently labeled low molecular weight, polar, non-toxic molecules; useful as aldehyde-fixable cell tracers
  • Membrane impermeant, can be easily loaded into cells by microinjection
  • Can be used to fluorescently label aldehyde or ketone groups of polysaccharides, carbohyrdates, or glycoproteins
  • Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent imaging
Bright, extremely water-soluble and non-toxic CF® Dye hydrazides are excellent fluorescent tracers for neurons or gap junction studies. The cell impermeant tracer dyes can be introduced into cells by microinjection.
CF® Dye hydrazides can also be used to fluorescently label aldehyde or ketone groups of carbohydrates or glycoproteins molecules after peroxidation with periodate.
For carbohydrates labeling applications, we recommend using CF® Dye aminooxy forms, which are more reactive toward these groups than hydrazide forms. For fluorescence two-dimensional differential gel electrophoresis (2D-DIGE), we recommend CFDI hydrazides.
Product Family Options
  • Désignation
    Ex/Em
    Taille
  • CF®350 hydrazide
    347/448 nm
    1 mg
    Each (1mg)
    Each (1mg) - $340.13
    $340.13
    Each (1mg)
       $340.13
     : 92151
  • CF®405S hydrazide
    404/431 nm
    1 mg
    Each
    Each - $348.40
    $348.40
    Each
       $348.40
     : 92183
  • CF®488A hydrazide
    490/515 nm
    1 mg
    Each (1mg)
    Each (1mg) - $374.19
    $374.19
    Each (1mg)
       $374.19
     : 92152
  • CF®555 hydrazide
    555/565 nm
    1 mg
    Each (1mg)
    Each (1mg) - $340.13
    $340.13
    Each (1mg)
       $340.13
     : 92153
  • CF®568 hydrazide
    562/583 nm
    1 mg
    Each (1mg)
    Each (1mg) - $340.13
    $340.13
    Each (1mg)
       $340.13
    Minimum order quantity is 1
     : 92154
  • CF®594 hydrazide
    593/614 nm
    1 mg
    Each (1mg)
    Each (1mg) - $340.13
    $340.13
    Each (1mg)
       $340.13
    Minimum order quantity is 1
     : 92158
  • CF®633 hydrazide
    630/650 nm
    1 mg
    Each (1mg)
    Each (1mg) - $340.13
    $340.13
    Each (1mg)
       $340.13
    Minimum order quantity is 1
     : 92156
  • CF®640R hydrazide
    642/662 nm
    1 mg
    Each (1mg)
    Each (1mg) - $340.13
    $340.13
    Each (1mg)
       $340.13
    Minimum order quantity is 1
     : 92157
  • CF®647 hydrazide
    650/665 nm
    1 mg
    Each (1mg)
    Each (1mg) - $374.19
    $374.19
    Each (1mg)
       $374.19
    Minimum order quantity is 1
     : 92136