Dilution Vials, Caplugs | Avantor
Dilution Vials, Caplugs
 :   Caplugs
Dilution Vials, Caplugs
 :  Caplugs
Dilution Vials, Caplugs
Dilution Vials, Caplugs
Dilution vials provide accurate hemolysis and distortion-free cell and particle counts.
  • Provided in natural color
  • One-piece polystyrene design includes rounded seams, a frosted writing area and optically clear parallel sides for optimum light transmission
  • Leakproof caps have convenient tabs for easy removal
Vials are available in cases of 5×50 tray packs or in bulk pack of 144.
Product Family Options
  • Capacité
    Matière
    Color
    Avec bouchon
    Cap Material
    Cap Color
    Packaging
  • 25.00 ml
    PS
    Natural
    Oui
    LDPE
    Natural
    5×50 tray packs
    Carton of 250
    Carton of 250 - $152.48
    $152.48
    Carton of 250
       $152.48
     : 201-5266-050
  • 25.00 ml
    PS
    Natural
    Oui
    LDPE
    Natural
    Bulk pack
    Carton of 144
    Carton of 144 - $79.67
    $79.67
    Carton of 144
       $79.67
     : 201-5266-05K