ContactUsLinkComponent
Free-Standing Light-Blocking Screw-Cap Microcentrifuge Tube, 2.0 ml
Catalog # 470335-056
Supplier:  MTC BIO, INC.
undefined
Free-Standing Light-Blocking Screw-Cap Microcentrifuge Tube, 2.0 ml
Catalog # 470335-056
Supplier:  MTC BIO, INC.
Supplier Number:  C2230-AM
Specifications
  • Volume:
    2.0 ml
  • Color:
    Amber
  • Description:
    SureSeal screw-cap tube
  • Cat. no.:
    470335-056
  • Maximum RCF:
    22000×g
Specifications
About this item
Free-standing microcentrifuge tube in light-blocking transparent amber color can hold up to 2.0 ml.
  • Made of ultra-clear polypropylene
  • Light-blocking amber color preserves integrity of sample
  • Leak-proof O-ring seal
  • RNase, DNase, and pyrogen free
  • Molded graduations
These tubes also come sterile and with caps assembled for quick and easy usage. May be centrifuged up to 22000×g.