Fischertechnik TXT 4.0 Base Set
Fischertechnik TXT 4.0 Base Set
Supplier:   STUDICA INC.
Fischertechnik TXT 4.0 Base Set
Supplier:  STUDICA INC.
Certificates
Fischertechnik TXT 4.0 Base Set
Fischertechnik TXT 4.0 Base Set
2 Options Available Below
Product Details & Documents
Measurement, control, regulation, and programming for middle/high school and beyond.
  • Acts as the core set for all fischertechnik robotic add-on products
  • Includes downloadable teaching and activity material (free download)
Main topics include robotics and programming (graphical and text-based), actuators, analog and digital sensors, measurement, control, data transmission, coding-decoding, and image processing. Set included a robotics TXT 4.0 controller, robo pro coding software, accu set, 2 encoder motors, USB camera, ultrasonic sensor, track sensor, 2 push button switches, 2 LEDs, and a photo-transistor.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Fischertechnik TXT 4.0 base set
    Discontinued
    Supplier Number: 559888
  • Fischertechnik TXT 4.0 controller only
    Log in for availability
    Log in for availability
    Each
    Each - $795.45
    $795.45
    Each
       $795.45
    Supplier Number: 560166
  • Fischertechnik TXT 4.0 omniwheels
    Log in for availability
    Log in for availability
    Each
    Each - $485.50
    $485.50
    Each
       $485.50
    Supplier Number: 559898