Extra Large Cutters, 7260, Excelta | Avantor
Extra Large Cutters, 7260, Excelta
Supplier:   Excelta
Extra Large Cutters, 7260, Excelta
Supplier:  Excelta
Certificates
Extra Large Cutters, 7260, Excelta
Extra Large Cutters, 7260, Excelta
1 Options Available Below
Product Details & Documents
The 7260 Series extra large oval cutter has a cutting capacity of .005" to .08" soft wire.
  • High grade carbon steel
  • Induction hardened edges
  • Can be re-sharpened
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Tip Size
    Availability
    Price
  • Cutters, very lage oval head, semi-flush
    Carbon
    5" (125 mm)
    0.035×0.035"
    Discontinued
    Supplier Number: 7260E
  • Cutters, very lage oval head, optimum flush
    Carbon
    5" (125 mm)
    0.035×0.035"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $176.89
    $176.89
    Each
       $176.89
    Supplier Number: 7261E