Miniature Wire Cutters, Excelta | Avantor
Miniature Wire Cutters, Excelta
Supplier:   Excelta
Miniature Wire Cutters, Excelta
Supplier:  Excelta
Certificates
Miniature Wire Cutters, Excelta
Miniature Wire Cutters, Excelta
6 Options Available Below
Product Details & Documents
These high-quality cutters are designed for cutting small soft wire up to 250,000 times without resharpening.
  • Three head shapes for various accessibility issues
  • Three types of cuts available
  • 'Set-Screw' stop prevents overtravel
  • Induction hardened blades for longevity
  • Ergonomic handles/dual leaf springs
These Lazer Line® side cutters are made of carbon tool steel with induction hardened cutting blades to RHC 63-65. The CNC machining allows the joint surfaces to be perfect to allow for long life. The dual leaf springs provide a smooth action. They feature an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. The application for these tools is soft wire and after up to 200,000 cuts, these tools can be factory resharpened.
Product Family Options
Oval head
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Curvature
    Availability
    Price
  • Cutters, small oval head, optimum flush
    Carbon
    4.25" (106.25 mm)
    14.25°
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $170.64
    $170.64
    Each
       $170.64
    Supplier Number: 9231E
  • Cutters, small oval head, optimum flush, slim line
    Carbon
    4" (100 mm)
    14.25°
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $170.64
    $170.64
    Each
       $170.64
    Supplier Number: 9231SL
  • Cutters, small oval head, laser flush
    Carbon
    4.25" (106.25 mm)
    14.25°
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $170.42
    $170.42
    Each
       $170.42
    Supplier Number: 9232E
  • Cutters, small oval head, laser flush, slim line
    Carbon
    4" (100 mm)
    14.25°
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $173.73
    $173.73
    Each
       $173.73
    Supplier Number: 9232SL
Tapered/relieved head
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Curvature
    Availability
    Price
  • Cutters, small taper relieved head, laser flush
    Carbon
    4.25" (106.25 mm)
    14.25°
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $182.31
    $182.31
    Each
       $182.31
    Supplier Number: 9238E
  • Cutters, small taper relieved head, laser flush, slim line
    Carbon
    4" (100 mm)
    14.25°
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $185.85
    $185.85
    Each
       $185.85
    Supplier Number: 9238SL