Super Spring with Stand | Avantor
Super Spring with Stand
Super Spring with Stand
Catalog # 470332-610
Fournisseur #:  SSPRING
Super Spring with Stand
Catalog # 470332-610
Super Spring with Stand
Super Spring with Stand
Super Spring with Stand
Liquidation
23,20 $
Vente
Valable jusqu'au 31/12/2026
Log in pour la disponibilité
Each
Product Details & Documents
Huge wave demonstrations.
  • Extra-long spring
  • Includes activity guide
Extra-long spring (21cm when compressed) allows users to magnify the wave demonstrations to illustrate traveling waves and wave patterns. Included stand prevents tangling or damage to super spring during storage.
Spécifications
  • Désignation:
    Super Spring with Stand