Super Spring with Stand
Super Spring with Stand
Catalog # 470332-610
Supplier: United Scientific Supplies
Fournisseur #: SSPRING
Jump to:
Product Details & Documents
Huge wave demonstrations.
- Extra-long spring
- Includes activity guide
Extra-long spring (21cm when compressed) allows users to magnify the wave demonstrations to illustrate traveling waves and wave patterns. Included stand prevents tangling or damage to super spring during storage.
Spécifications
- Désignation:Super Spring with Stand