Dual Conductor Speidel Metal Wrist Strap, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
The Speidel metal band features two 180 degree conductive paths around the wrist, providing maximum skin contact.
- Premium metal expansion wrist strap with more reliable path-to-ground
- Durable metal band is designed for long wearing, and is easy to clean
- Dual-Wire conductor provides redundancy
- If one dual-wire wrist strap wire is severed, operator has reliable path-to-ground with other wire
The expandable band is constructed with two stainless steel back plates and is available in small, medium, and large sizes.
Frequently Bought Together