ContactUsLinkComponent
Air Track Gliders
Catalogue # 470224-382
Fournisseur:  Wards
undefined
Air Track Gliders
Catalogue # 470224-382
Fournisseur:  Wards
Supplier Number:  PH0362GLD2
Spécifications
  • Description:
    Air track gliders
  • Cat. no.:
    470224-382
  • Supplier No.:
    PH0362GLD2
Spécifications
À propos de cet article
A pair of 100g gliders designed for use with air tracks 470157-654 and 470157-664.
  • Made from anodized aluminum
  • Each glider has flanks that are perpendicular to each other and sit 45 degrees below the horizontal
  • Trails along the side of the glider allow for the attachment of masses, flags, or other objects
  • Gliders can be used to simulate motion and kinematics in a near-frictionless environment on the air track
High quality 100g gliders designed with unique features including the ability to add masses, flags, or other objects. Compatible with a variety of air tracks similar to 470157-654 and 470157-664.