Buffers for E.Z.N.A.® Gel Extraction Systems, Omega Bio-tek®
Supplier: Omega Bio-Tek
Certificates
Certificates
About this item
Frequently Bought Together
Parafilm® M Sealing Film
Supplier: Heathrow Scientific
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
Acétone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
CHEMets® Visual Kit, Hydrogen Peroxide Test Kit, CHEMetrics
Supplier: CHEMetrics
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
Propanol-2 ≥99.8% (by GC), HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Masterflex® L/S® High-Performance Precision Pump Tubing, Versilon™ A-60-G, Avantor®
Supplier: VWR International
SOLUTION RNASE-X DECONTAMINATION 250ML
Supplier: Biotium
Dissection Tool with Dual Prongs, Mortech
Supplier: Mortech Manufacturing
Human Recombinant CNTF (from <i>E. coli</i>)
Supplier: Peprotech
DAPT ≥98% (by NMR)
Supplier: Enzo Life Sciences
Phosphate Buffer
Supplier: Ward's Science