MXP Data Loggers, Cryopak Verification Technologies
MXP Data Loggers, Cryopak Verification Technologies
MXP Data Loggers, Cryopak Verification Technologies
Certificates
MXP Data Loggers, Cryopak Verification Technologies
MXP Data Loggers, Cryopak Verification Technologies
4 Options Available Below
Product Details & Documents
The MXP data logger offers flexibility and longevity with a light sensor to detect and log if package is opened.
  • Long shelf life
  • Event logging feature
  • Internal digital temperature sensor
With its USB connectivity, the MXP data logger provides detailed reports in both PDF and Excel format, and is user friendly for both Windows and smartphone app use.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Data logger, MXP single use
    Log in for availability
    Log in for availability
    Case of 25
    Case of 25 - $2,078.07
    $2,078.07
    Case of 25
       $2,078.07
    Supplier Number: MXP-S
  • Data logger, MXP single use dry ice
    Discontinued
    Supplier Number: -
  • Data logger, MXP multi use
    Log in for availability
    Log in for availability
    Each
    Each - $122.04
    $122.04
    Each
       $122.04
    Supplier Number: MXP-M
  • Data logger, MXP multi use dry ice
    Log in for availability
    Log in for availability
    Each
    Each - $175.78
    $175.78
    Each
       $175.78
    Supplier Number: MXPD-M
  • Data logger, MXP multi use with RF on
    Log in for availability
    Log in for availability
    Each
    Each - $154.67
    $154.67
    Each
       $154.67
    Supplier Number: MXP-RAO