Raised Panel Polyester Rectangular Labels, 3" Core, Brady
Raised Panel Polyester Rectangular Labels, 3" Core, Brady
Supplier:   Brady Worldwide
Raised Panel Polyester Rectangular Labels, 3" Core, Brady
Supplier:  Brady Worldwide
Certificates
Raised Panel Polyester Rectangular Labels, 3" Core, Brady
Raised Panel Polyester Rectangular Labels, 3" Core, Brady
3 Options Available Below
Product Details & Documents
Raised panel Adhesive-Taped Polyester (B-593) labels are designed for patch panel identification, external push-buttons, switches, and internal connection points.
  • Raised panel design
  • Aggressive adhesive
  • Solvent-resistant
Raised panel Adhesive-Taped Polyester (B-593) labels can be used as rating and serial plates using alphanumerics that require nameplate quality. Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags. Service temperature of ‒20 to +100 °C (‒4 to +212 °F)
Labels are designed for use with BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, THT 3" Core Printers.
Certifications: UL approved.
Ordering information: A ribbon is required to print this material. Use Part Number 49006-110 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use R6006 for a different printer.
Product Family Options
Product Information
  • Désignation
    Matière
    Color
    Core Size
    Dimensions
    Packaging
    Availability
    Price
  • Raised panel labels
    Polyester
    White
    3"
    12.4×26.9 mm (0.49×16")
    500 labels/roll
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $497.50
    $497.50
    New Customer Promotional Price
    Roll of 1
       $497.50
    Supplier Number: THTEP-171-593-.5
  • Raised panel labels
    Polyester
    White
    3"
    25.4×50.8 mm (1×2")
    500 labels/roll
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $590.03
    $590.03
    New Customer Promotional Price
    Roll of 1
       $590.03
    Supplier Number: THTEP-173-593-.5
  • Raised panel labels
    Polyester
    White
    3"
    25.4×76.2 mm (1×3")
    500 labels/roll
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $1,076.70
    $1,076.70
    New Customer Promotional Price
    Roll of 1
       $1,076.70
    Supplier Number: THTEP-176-593-.5