Metallized High Adhesion Matte Polyester Labels, 3" Core, Brady
Metallized High Adhesion Matte Polyester Labels, 3" Core, Brady
Supplier:   Brady Worldwide
Metallized High Adhesion Matte Polyester Labels, 3" Core, Brady
Supplier:  Brady Worldwide
Certificates
Metallized High Adhesion Matte Polyester Labels, 3" Core, Brady
Metallized High Adhesion Matte Polyester Labels, 3" Core, Brady
3 Options Available Below
Product Details & Documents
Ultra-aggressive metallized polyester (B-486B) is ideal for rating/serial plates that use barcodes, alphanumerics, graphic symbols, and logos, and require nameplate-like quality.
  • Aggressive, heat-resistant, permanent adhesive
  • Service temperature: ‒40 to +120 °C (‒40 to +248 °F)
Matte, light gray material has a permanent rubber-based adhesive and is thermal transfer printable. Designed for high adhesion to textured metals, powder-coated surfaces, and low energy plastics.
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, or THT 3" Core Printers.
 : A ribbon is required to print this material. Use Part Number 89077-374 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use 94000-380 for a different printer.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Color
    Core Size
    Dimensions
    Packaging
    Availability
    Price
  • High adhesion labels, metallized with matte finish
    Polyester
    Light grey
    3"
    25.4×50.8 mm (1×2")
    3000 labels/roll
    Log in pour la disponibilité
    Log in pour la disponibilité
    Roll of 1
    Roll of 1 - $663.64
    $663.64
    Prix promotionnel pour les nouveaux clients
    Roll of 1
       $663.64
    Supplier Number: THT-17-486-3
  • High adhesion labels, metallized with matte finish
    Polyester
    Light grey
    3"
    25.4×76.2 mm (1×3")
    3000 labels/roll
    Log in pour la disponibilité
    Log in pour la disponibilité
    Roll of 1
    Roll of 1 - $929.74
    $929.74
    Prix promotionnel pour les nouveaux clients
    Roll of 1
       $929.74
    Supplier Number: THT-18-486-3
  • High adhesion labels, metallized with matte finish
    Polyester
    Light grey
    3"
    50.8×76.2 mm (2×3")
    1000 labels/roll
    Log in pour la disponibilité
    Log in pour la disponibilité
    Roll of 1
    Roll of 1 - $638.70
    $638.70
    Prix promotionnel pour les nouveaux clients
    Roll of 1
       $638.70
    Supplier Number: THT-19-486-1