UV Indicator Stickers
Uv indicator stickers
Catalog # 470334-104
Supplier: Ward's
Fournisseur #: B1450-ISS
Jump to:
Product Details & Documents
Indicator cards and stickers allow for quick determination of UV-C exposure.
- Color change clearly indicates UV exposure
- Convenient sticker form
- Pack of 40
Each sticker measures 0.5" square.
Spécifications
- Désignation:Uv indicator stickers