Purospher® HPLC Columns, Agilent Technologies
Catalog # CA79925PU-564
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Column Format:Analytical
- Ø int.:4.00 mm
- Particle Size:5 µm
- Pore size:80 Å
- Length:125 mm
- Cat. no.:CA79925PU-564
- Phase:RP-18
- Supplier No.:79925PU-564