Axygen® Tip Refill System, Empty Racks, Corning | Avantor
Axygen® Tip Refill System, Empty Racks, Corning
Supplier:   Corning
Axygen® Tip Refill System, Empty Racks, Corning
Supplier:  Corning
Certificates
Axygen® Tip Refill System, Empty Racks, Corning
2 Options Available Below
Product Details & Documents
Axygen® Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable.
  • RFL-R-1200 is suitable for 1200 µl refill inserts
  • RFL-R is suitable for 10 and 200 µl refill inserts
  • RNase- and DNase-free, non pyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
They are available in different colors and are packaged in bulk or racks.
Product Family Options
Informations sur le produit
  • Désignation
    For Use With
    Availability
    Price
  • Empty racks; 10 racks/pack, 5 packs/case
    10, 200 µl Axygen® tips
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $35.60
    $35.60
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $178.00
    $178.00
    Pack of 10
       $35.60
    Case of 50
       $178.00
    Supplier Number: RFL-R
  • Empty racks; 10 racks/pack, 3 packs/case
    1000, 1200 µl Axygen® tips
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $44.40
    $44.40
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $133.20
    $133.20
    Prix promotionnel pour les nouveaux clients
    Pack of 10
       $44.40
    Case of 30
       $133.20
    Supplier Number: RFL-R-1200