Scott® Essential Coreless Standard Roll Bathroom Tissue, Kimberly-Clark Professional
Catalog # CA04007
Supplier: Kimberly-Clark
Specifications
- Color:White
- Description:Coreless standard bath tissue roll
- Cat. no.:CA04007
- Supplier No.:04007
Specifications
About this item
Scott® Essential provides reliable balance of efficiency, performance, and value. Scott® Essential toilet paper provides any office a reliable washroom experience and allows fewer refills, less waste, and easy maintenance.
- 2-Ply Scott® Essential coreless toilet paper is efficient with fewer refills, less waste, and easy maintenance
- Convenient storage and product handling
- Each roll is nearly 50% longer than standard roll bathroom tissue with 100% core and paper wrap elimination
- Can be used with the coreless Kimberly-Clark Professional toilet paper commercial dispenser
As a coreless bath tissue with high capacity features, each roll is nearly 50% longer than standard roll bath tissue. As an environmentally friendly toilet tissue, it eliminates the cardboard core and paper wrap, meets EPA standards, and is FSC® and Ecologo® certified. It can be used in most coreless toilet paper commercial dispensers.
Certifications: FSC® and Ecologo® certified. Meets or exceeds EPA guidelines for post-consumer waste content (at least 25%).
Packaging: 36 rolls per case, 1000 sheets per roll, 36000 sheets per case.