Transport Vial Caps, Caplugs
Supplier: Caplugs
Certificates
About this item
Caplugs Transport Vial Caps are designed to fit all sizes of transport vials.
- Caps also fit 15 ml threaded centrifuge tubes
- All transport vials are 95 kPa compliant
Available in a wide range of colors.
Frequently Bought Together