VWR® Plus Series Refrigerator Freezer Combo Units with Natural Refrigerants
Supplier: VWR International
Certificates
About this item
The joint refrigerator and freezer combo units provide quality cold storage for laboratory settings that require both frozen and refrigerated materials.
- Microprocessor temperature controller
- Digital temperature display
- Audible and visual high and low temperature alarms
- Remote alarm contacts
- Keyed door lock
- Probe access port (⅜")
- HFC-free refrigerant (R600a)
These economical combo units offer a cold-storage solution for both refrigerated and frozen storage in laboratory settings. These units are equipped with premium features including a series of temperature alarms to keep stored products at peak effectiveness, keyed door locks, LED interior lighting, a probe access port, and a microprocessor temperature controller.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty. They run on HFC-free and hydrocarbon refrigerant for environmental cleanliness and economic efficiency.
Certifications: UL/C-UL listed
Caution: *Compatible with refrigerator compartment only.
Specifications
- Catalog No:
- 76488-184
- 76488-186
- 76488-188
- 76488-190
- 76488-192
- 76488-178
- 76488-180
- 76488-182
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76488-184
- 76488-186
- 76488-188
- 76488-190
- 76488-192
- 76488-178
- 76488-180
- 76488-182
- Defrost:
- Manual defrost (freezer); Cycle defrost (refrigerator)
- Manual defrost (freezer); Cycle defrost (refrigerator)
- Manual defrost (freezer); Cycle defrost (refrigerator)
- Manual defrost (freezer); Cycle defrost (refrigerator)
- Auto defrost (freezer); Cycle defrost (refrigerator)
- Manual defrost (freezer); Cycle defrost (refrigerator)
- Manual defrost (freezer); Cycle defrost (refrigerator)
- Manual defrost (freezer); Cycle defrost (refrigerator)
- Shelves/drawers:
- 2 Fixed shelves (Freezer); 3 Adjustable shelves (Refrigerator)
- 3 Adjustable shelves (Freezer); 3 Adjustable shelves (Refrigerator)
- 3 Adjustable shelves (Freezer); 3 Adjustable shelves (Refrigerator)
- 2 Adjustable shelves (Freezer); 3 Adjustable shelves (Refrigerator)
- 2 Adjustable shelves (Freezer); 3 Adjustable shelves (Refrigerator)
- 2 Fixed shelves (Freezer); 3 Adjustable shelves (Refrigerator)
- 3 Adjustable shelves (Freezer); 3 Adjustable shelves (Refrigerator)
- 3 Adjustable shelves (Freezer); 3 Adjustable shelves (Refrigerator)
- Door type:
- One glass swing door (refrigerator); One solid swing door (freezer)
- One glass swing door (refrigerator); One solid swing door (freezer)
- One glass swing door (refrigerator); One solid swing door (freezer)
- One solid swing door (refrigerator); One solid swing door (freezer)
- One solid swing door (refrigerator); One solid swing door (freezer)
- One solid swing door (refrigerator); One solid swing door (freezer)
- One solid swing door (refrigerator); One solid swing door (freezer)
- One solid swing door (refrigerator); One solid swing door (freezer)
- Specifications:
- White cabinet and door
- White cabinet and door
- White cabinet and door
- White powder coated exterior
- White powder coated exterior
- White cabinet and door
- White cabinet and door
- White cabinet and door
- Electrical:
- 115V, 60 Hz, 1.4 Amps, 1/8 HP (Freezer); 1.3 Amps, 1/6 HP (Refrigerator)
- 115V, 60 Hz, 2.2 Amps, 1/4 HP (Freezer); 1.3 Amps, 1/6 HP (Refrigerator)
- 115V, 60 Hz, 2.2 Amps, 1/4 HP (Freezer); 1.3 Amps, 1/6 HP (Refrigerator)
- 115V, 60 Hz, 1 Amp, 1/8 HP (Freezer); 1.3 Amps, 1/6 HP (Refrigerator)
- 115V, 60 Hz, 1 Amp, 1/8 HP (Freezer); 1.3 Amps, 1/6 HP (Refrigerator)
- 115V, 60 Hz, 1.4 Amps; 1/8 HP (Freezer); 1.6 Amps, 1/6 HP (Refrigerator)
- 115V, 60 Hz, 2.2 Amps, 1/4 HP (Freezer); 1.3 Amps, 1/6 HP (Refrigerator)
- 115V, 60 Hz, 2.2 Amps, 1/4 HP (Freezer); 1.3 Amps, 1/6 HP (Refrigerator)
- Weight:
- 121 kg (266 lbs.)
- 122 kg (268 lbs.)
- 122 kg (268 lbs.)
- 98 kg (215 lbs.)
- 98 kg (215 lbs.)
- 121 kg (266 lbs.)
- 122 kg (268 lbs.)
- 122 kg (268 lbs.)