ContactUsLinkComponent
Scott® Pro Liquid Hand Soap with Moisturizers, Kimberly-Clark Professional
Catalogue # 76333-054
Fournisseur:  Kimberly-Clark
undefined
Scott® Pro Liquid Hand Soap with Moisturizers, Kimberly-Clark Professional
Catalogue # 76333-054
Fournisseur:  Kimberly-Clark
Supplier Number:  91592
Spécifications
  • Capacity:
    1200 ml (40.6 oz.)
  • Color:
    Pink
  • Description:
    Liquid hand soap with moisturizers, floral scent
  • Cat. no.:
    76333-054
Spécifications
À propos de cet article
Scott® Pro Liquid Hand Soap is a luxurious, moisturizing pink foam soap, featuring a pleasant floral scent. Same Kleenex® quality, now Scott® branded.
  • Scott® Pro foaming soap is an important step in the washroom experience and ideal for use in commercial, high-traffic areas
  • High-capacity, wall-mount container provides long-lasting use, reducing maintenance time and costs
  • Use with a compatible Kimberly-Clark Professional dispenser for consistent dispensing
  • The easy-to-load bulk cassette clicks right into place, simplifying maintenance
Providing the right hygienic supplies in the restroom is a priority for businesses, showing staff and visitors that you care enough to provide what they need. Provide Scott® Pro Liquid Hand Soap to help stop the spread of germs. This luxurious, moisturizing pink liquid foam soap has a subtle floral scent and comes in a sealed container. Because the dispenser is high-capacity, it’s a popular choice for use in washrooms in office buildings, healthcare environments, high-traffic areas, hospitality, lodging, and commercial spaces. The easy-to-load cassette is designed to be compatible with Kimberly-Clark dispensers, and the bottle is recyclable (after the pump is removed).
Packaging: Two cassettes per case.