SP Bel-Art 96 Deep-Well Plates, Automation Compliant, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Used with multi-channel pipettes or with automation systems, these 96 Deep-Well Plates are made of virgin polypropylene.
- SBS/ANSI/SLAS format compatible
- DNase, RNase, pyrogen free, manufactured in Class 8 clean room
- Alphanumeric marking and cut-angle mark for easy referencing
- Can be used with multi-channel pipettes or automation systems
- Can be stored at –80 °C
- Autoclavable up to 121 °C
V-bottom shape is ideal for ease of sample retrieval. U-bottom shape is ideal for agglutination or applications where light stirring is necessary.
Frequently Bought Together