NEBNext® Ultra II RNA Library Prep Kit for Illumina, New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
The Ultra II RNA Library Prep Kit for Illumina is designed for non-directional (non-strand-specific) RNA library construction, and delivers significantly increased sensitivity and specificity from your RNA-seq experiments, from ever-decreasing amounts of input RNA.
- Generate the highest yields of high quality libraries, with a broad range of input amounts
- Increase the complexity and transcript coverage of your libraries
- Optimize your time with streamlined workflows, reduced hands-on time, and automation compatibility
- Rely on robust performance, even with low quality RNA, including FFPE
In conjunction with ribosomal RNA (rRNA) depletion or poly(A) mRNA enrichment, the kit enables the production of high quality libraries from 5 ng or 10 ng of Total RNA, respectively, up to 1 µg.
Frequently Bought Together