NEBNext® Multiplex Oligos for Illumina (Index Primers Set 4), New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.
- High efficiency adaptor ligation
- Minimized adaptor-dimer formation
- Includes index primers for library multiplexing
- Increased sample identification specificity
- Includes USER Enzyme
Frequently Bought Together