NEBNext® Multiplex Oligos for Illumina (Index Primers Set 3), New England Biolabs
Fournisseur: New England Biolabs (NEB)
Certificats
À propos de cet article
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.
- High efficiency adaptor ligation
- Minimized adaptor-dimer formation
- Includes index primers for library multiplexing
- Increased sample identification specificity
- Includes USER Enzyme
Frequently Bought Together