NEBNext® Ultra II DNA Library Prep with Sample Purification Beads, New England Biolabs
The NEBNext Ultra II DNA Library Prep with Sample Purification Beads contains the enzymes, buffers and purification beads required to convert a broad range of DNA input amounts into indexed libraries for next-generation sequencing on the Illumina platform.
- Get more of what you need, with the highest library yields
- Use to generate high quality libraries from a broad range of input amounts, from 500 pg to 1 µg
- Prepare libraries from ALL your samples, including GC-rich DNA and FFPE DNA samples
- Improve yields and quality for target enrichment applications
- Includes SPRIselect beads
The fast, user-friendly workflow has minimal hands-on time and, for added convenience, this kit contains NEBNext Sample Purification Beads (SPRIselect beads from Beckman Coulter) for size selection and enzyme reaction cleanup.
Frequently Bought Together