Immobilized T4 DNA Ligase, New England Biolabs
Catalog # CA76486-074
Supplier: New England Biolabs (NEB)
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Immobilized T4 DNA Ligase 10 mg/ml
- Size:1.1 mg
- Cat. no.:CA76486-074
Specifications
About this item
Immobilized T4 DNA Ligase (IM-T4 DNA Ligase) is a slurry of magnetic beads coated with T4 DNA Ligase to produce a 10 mg per ml solution (50% glycerol) with an effective concentration of 60 cohesive end units (CEU) per microliter of slurry. Following a reaction, enzyme can be removed using a magnet thereby avoiding heat inactivation, and can be re-used.
- Easily remove enzyme from reaction using a magnet
- Avoid heat inactivation step, which may interfere with downstream applications
- Reusable up to 20 times
- Enable new workflows and applications