ContactUsLinkComponent
HardyDisk™ AST Doripenem, 10 µg (DOR-10), Doribax, Hardy Diagnostics
Catalogue # 76334-642
Fournisseur:  Hardy Diagnostics
Numéro CAS:  
undefined
HardyDisk™ AST Doripenem, 10 µg (DOR-10), Doribax, Hardy Diagnostics
Catalogue # 76334-642
Fournisseur:  Hardy Diagnostics
Supplier Number:  Z9371
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Pack type:
    1×50 disks
  • Description:
    Test disks
  • Cat. no.:
    76334-642
Spécifications
À propos de cet article
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
  • Improved cartridge design helps to prevent jamming and tearing
  • HardyDisk™ AST cartridges are packaged in an all new canister that features a desiccant built into the lid
  • The last disk in each cartridge is clearly marked with dark lines around its edge to signify the need for refill
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.